Assay Details
| Gene Symbol: Coa8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272218.1 | NP_001259147.1 | 2 - 3 | 360 | 73 | |
| NM_130600.2 | NP_569956.1 | 2 - 3 | 609 | 73 | ||
More Information
Set Membership: |
|

Coa8
Genomic Map