Assay Details
| Gene Symbol: stum | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274196.2 | NP_001261125.2 | 4 - 5 | 4870 | 93 | |
| NM_137759.5 | NP_611603.3 | 5 - 6 | 5016 | 93 | ||
More Information
Set Membership: |
|

stum
Genomic Map