Assay Details
| Gene Symbol: stum | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274196.2 | NP_001261125.2 | 5 - 6 | 5511 | 64 | |
| NM_001316430.1 | NP_001303359.1 | 2 - 3 | 868 | 64 | ||
| NM_137759.5 | NP_611603.3 | 6 - 7 | 5657 | 64 | ||
More Information
Set Membership: |
|

stum
Genomic Map