Assay Details
| Gene Symbol: Gmer | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137890.3 | NP_611734.1 | 1 - 2 | 230 | 78 | |
| GenBank mRNA | AM231688.1 | - | 1 - 2 | 132 | 78 | |
| BT050562.1 | - | 1 - 2 | 225 | 78 | ||
More Information
Set Membership: |
|

Gmer
Genomic Map