Assay Details
| Gene Symbol: Gmer | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137890.3 | NP_611734.1 | 3 - 4 | 898 | 61 | |
| GenBank mRNA | AM231688.1 | - | 3 - 4 | 800 | 61 | |
| BT050562.1 | - | 3 - 4 | 893 | 61 | ||
More Information
Set Membership: |
|

Gmer
Genomic Map