Assay Details
| Gene Symbol: mr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138018.3 | NP_611862.1 | 7 - 8 | 2281 | 76 | |
| GenBank mRNA | AY129297.1 | - | 7 - 8 | 2203 | 76 | |
| BT031326.1 | - | 7 - 8 | 2248 | 76 | ||
More Information
Set Membership: |
|

mr
Genomic Map