Assay Details
| Gene Symbol: ocm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202069.1 | NP_001188998.1 | 4 - 5 | 2664 | 66 | |
| NM_001299902.1 | NP_001286831.1 | 3 - 4 | 2570 | 66 | ||
| NM_138050.4 | NP_611894.1 | 4 - 5 | 2837 | 66 | ||
More Information
Set Membership: |
|

ocm
Genomic Map