Assay Details
| Gene Symbol: ocm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202069.1 | NP_001188998.1 | 5 - 6 | 6574 | 81 | |
| NM_001299902.1 | NP_001286831.1 | 4 - 5 | 6480 | 81 | ||
| NM_138050.4 | NP_611894.1 | 5 - 6 | 6747 | 81 | ||
More Information
Set Membership: |
|

ocm
Genomic Map