Assay Details
| Gene Symbol: Ccs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169637.3 | NP_001163108.1 | 2 - 3 | 709 | 59 | |
| NM_143772.5 | NP_652029.3 | 2 - 3 | 691 | 59 | ||
More Information
Set Membership: |
|

Ccs
Genomic Map