Assay Details
| Gene Symbol: Usp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274299.1 | NP_001261228.1 | 4 - 5 | 2447 | 58 | |
| NM_001274300.1 | NP_001261229.1 | 7 - 8 | 3997 | 58 | ||
| NM_167844.2 | NP_728554.1 | 7 - 8 | 4263 | 58 | ||
| GenBank mRNA | AY122170.1 | - | 7 - 8 | 4251 | 58 | |
More Information
Set Membership: |
|

Usp10
Genomic Map