Assay Details
| Gene Symbol: hfp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169844.2 | NP_001163315.1 | 2 - 3 | 2973 | 75 | |
| NM_001169845.2 | NP_001163316.1 | 4 - 5 | 559 | 75 | ||
| NM_001169846.2 | NP_001163317.1 | 5 - 6 | 893 | 75 | ||
| NM_001169847.2 | NP_001163318.1 | 3 - 4 | 2572 | 75 | ||
| NM_001169848.2 | NP_001163319.1 | 4 - 5 | 1198 | 75 | ||
| NM_001169849.2 | NP_001163320.1 | 4 - 5 | 564 | 75 | ||
| NM_080384.4 | NP_525123.2 | 3 - 4 | 512 | 75 | ||
| NM_167880.3 | NP_728610.1 | 4 - 5 | 620 | 75 | ||
| NM_167881.3 | NP_728611.1 | 4 - 5 | 573 | 75 | ||
| NM_167882.3 | NP_728612.1 | 6 - 7 | 820 | 75 | ||
| GenBank mRNA | AY061006.1 | - | 6 - 7 | 819 | 75 | |
| BT003317.1 | - | 2 - 3 | 2989 | 75 | ||
More Information
Set Membership: |
|

hfp
Genomic Map