Assay Details
| Gene Symbol: Nf-YC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132126.3 | NP_572354.1 | 1 - 2 | 238 | 67 | |
| GenBank mRNA | BT001649.1 | - | 1 - 2 | 234 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nf-YC| Gene Symbol: Nf-YC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132126.3 | NP_572354.1 | 1 - 2 | 238 | 67 | |
| GenBank mRNA | BT001649.1 | - | 1 - 2 | 234 | 67 | |
Set Membership: |
|