Assay Details
| Gene Symbol: Nf-YC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132126.3 | NP_572354.1 | 2 - 3 | 1605 | 84 | |
| GenBank mRNA | BT001649.1 | - | 2 - 3 | 1601 | 84 | |
More Information
Set Membership: |
|

Nf-YC
Genomic Map