Assay Details
| Gene Symbol: Rab39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258639.2 | NP_001245568.1 | 1 - 2 | 414 | 65 | |
| NM_132177.4 | NP_572405.2 | 1 - 2 | 414 | 65 | ||
| GenBank mRNA | BT044331.1 | - | 1 - 2 | 412 | 65 | |
More Information
Set Membership: |
|

Rab39
Genomic Map