Assay Details
| Gene Symbol: Upf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132206.3 | NP_572434.1 | 2 - 3 | 3258 | 79 | |
| GenBank mRNA | BT024221.1 | - | 2 - 3 | 3234 | 79 | |
More Information
Set Membership: |
|

Upf2
Genomic Map