Assay Details
| Gene Symbol: Nost | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169240.2 | NP_001162711.1 | 6 - 7 | 3042 | 77 | |
| NM_001169241.1 | NP_001162712.1 | 6 - 7 | 1634 | 77 | ||
| NM_001201637.1 | NP_001188566.1 | 6 - 7 | 2140 | 77 | ||
| NM_001201638.2 | NP_001188567.1 | 6 - 7 | 3003 | 77 | ||
| NM_001298100.1 | NP_001285029.1 | 7 - 8 | 3826 | 77 | ||
| NM_132287.3 | NP_572515.2 | 5 - 6 | 1861 | 77 | ||
| GenBank mRNA | BT044544.1 | - | 6 - 7 | 3042 | 77 | |
| BT124862.1 | - | 6 - 7 | 1634 | 77 | ||
| BT133107.1 | - | 6 - 7 | 2141 | 77 | ||
More Information
Set Membership: |
|

Nost
Genomic Map