Assay Details
| Gene Symbol: Oseg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274349.1 | NP_001261278.1 | 2 - 3 | 319 | 68 | |
| NM_139396.4 | NP_647653.1 | 2 - 3 | 319 | 68 | ||
| GenBank mRNA | AY061373.1 | - | 2 - 3 | 178 | 68 | |
More Information
Set Membership: |
|

Oseg4
Genomic Map