Assay Details
| Gene Symbol: ProRS-m | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139480.3 | NP_647737.1 | 2 - 3 | 447 | 70 | |
| GenBank mRNA | BT150200.1 | - | 2 - 3 | 384 | 70 | |
More Information
Set Membership: |
|

ProRS-m
Genomic Map