Assay Details
| Gene Symbol: CG32371 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_168231.2 | NP_729295.1 | 1 | 926 | 139 | |
| GenBank mRNA | BT081980.1 | - | 1 | 903 | 139 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CG32371| Gene Symbol: CG32371 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_168231.2 | NP_729295.1 | 1 | 926 | 139 | |
| GenBank mRNA | BT081980.1 | - | 1 | 903 | 139 | |
Set Membership: |
|