Assay Details
| Gene Symbol: Nrx-IV | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_168491.3 | NP_729787.2 | 2 - 3 | 562 | 73 | |
| GenBank mRNA | BT120004.1 | - | 2 - 3 | 529 | 73 | |
More Information
Set Membership: |
|

Nrx-IV
Genomic Map