Assay Details
| Gene Symbol: futsch | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103394.3 | NP_001096864.1 | 7 - 8 | 16783 | 67 | |
| NM_001169161.3 | NP_001162632.2 | 8 - 9 | 17419 | 67 | ||
| NM_001258532.1 | NP_001245461.1 | 8 - 9 | 18219 | 67 | ||
More Information
Set Membership: |
|

futsch
Genomic Map