Assay Details
| Gene Symbol: Upf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298216.1 | NP_001285145.1 | 2 - 3 | 543 | 68 | |
| NM_132539.3 | NP_572767.1 | 2 - 3 | 572 | 68 | ||
| GenBank mRNA | AY061379.1 | - | 2 - 3 | 541 | 68 | |
More Information
Set Membership: |
|

Upf1
Genomic Map