Assay Details
| Gene Symbol: mRpL49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298241.1 | NP_001285170.1 | 1 - 2 | 140 | 57 | |
| NM_132611.3 | NP_572839.1 | 1 - 2 | 140 | 57 | ||
| GenBank mRNA | BT126389.1 | - | 1 - 2 | 135 | 57 | |
More Information
Set Membership: |
|

mRpL49
Genomic Map