Assay Details
| Gene Symbol: DNAlig4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132679.3 | NP_572907.1 | 3 - 4 | 1456 | 56 | |
| GenBank mRNA | BT057979.1 | - | 3 - 4 | 1454 | 56 | |
More Information
Set Membership: |
|

DNAlig4
Genomic Map