Assay Details
| Gene Symbol: Fbxl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132723.3 | NP_572951.1 | 8 - 9 | 2474 | 62 | |
| GenBank mRNA | AY069093.1 | - | 8 - 9 | 2453 | 62 | |
More Information
Set Membership: |
|

Fbxl4
Genomic Map