Assay Details
| Gene Symbol: PPYR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132784.2 | NP_573012.1 | 1 | 927 | 129 | |
| GenBank mRNA | AY118699.1 | - | 1 | 927 | 129 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PPYR1| Gene Symbol: PPYR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132784.2 | NP_573012.1 | 1 | 927 | 129 | |
| GenBank mRNA | AY118699.1 | - | 1 | 927 | 129 | |
Set Membership: |
|