Assay Details
| Gene Symbol: CG9784 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272694.2 | NP_001259623.1 | 1 | 1879 | 61 | |
| NM_001272695.2 | NP_001259624.1 | 2 | 1875 | 61 | ||
| NM_132907.3 | NP_573135.1 | 1 | 1879 | 61 | ||
| GenBank mRNA | AY119249.1 | - | 1 | 1853 | 61 | |
More Information
Set Membership: |
|

CG9784
Genomic Map