Assay Details
| Gene Symbol: CG7376 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274542.1 | NP_001261471.1 | 2 - 3 | 1352 | 60 | |
| NM_139777.2 | NP_648034.1 | 2 - 3 | 1352 | 60 | ||
More Information
Set Membership: |
|

CG7376
Genomic Map