Assay Details
| Gene Symbol: Cdc27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274574.1 | NP_001261503.1 | 2 - 3 | 762 | 80 | |
| NM_139836.3 | NP_648093.2 | 2 - 3 | 762 | 80 | ||
| GenBank mRNA | BT021947.1 | - | 2 - 3 | 739 | 80 | |
More Information
Set Membership: |
|

Cdc27
Genomic Map