Assay Details
| Gene Symbol: Oseg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300053.1 | NP_001286982.1 | 11 - 12 | 4567 | 66 | |
| NM_139964.3 | NP_648221.3 | 10 - 11 | 3515 | 66 | ||
| GenBank mRNA | BT053694.1 | - | 10 - 11 | 3496 | 66 | |
More Information
Set Membership: |
|

Oseg1
Genomic Map