Assay Details
| Gene Symbol: Oseg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300053.1 | NP_001286982.1 | 9 - 10 | 4128 | 100 | |
| NM_139964.3 | NP_648221.3 | 8 - 9 | 3076 | 100 | ||
| GenBank mRNA | BT053694.1 | - | 8 - 9 | 3057 | 100 | |
More Information
Set Membership: |
|

Oseg1
Genomic Map