Assay Details
| Gene Symbol: RpLP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272893.1 | NP_001259822.1 | 1 - 2 | 195 | 68 | |
| NM_057282.4 | NP_476630.1 | 1 - 2 | 195 | 68 | ||
| GenBank mRNA | AY069125.1 | - | 1 - 2 | 216 | 68 | |
| S62170.1 | - | 1 - 2 | 79 | 68 | ||
More Information
Set Membership: |
|

RpLP1
Genomic Map