Assay Details
| Gene Symbol: so | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057385.5 | NP_476733.1 | 4 - 5 | 1475 | 75 | |
| GenBank mRNA | BT029256.1 | - | 4 - 5 | 1459 | 75 | |
| L31626.1 | - | 4 - 5 | 1791 | 75 | ||
More Information
Set Membership: |
|

so
Genomic Map