Assay Details
| Gene Symbol: aub | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103674.3 | NP_001097144.1 | 1 - 2 | 320 | 88 | |
| NM_057386.5 | NP_476734.1 | 2 - 3 | 337 | 88 | ||
| GenBank mRNA | AF145680.1 | - | 2 - 3 | 254 | 88 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001103674.3 transcripts.
Set Membership: |
|

aub
Genomic Map