Assay Details
| Gene Symbol: aub | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103674.3 | NP_001097144.1 | 2 - 3 | 813 | 69 | |
| NM_057386.5 | NP_476734.1 | 3 - 4 | 830 | 69 | ||
| GenBank mRNA | AF145680.1 | - | 3 - 4 | 747 | 69 | |
More Information
Set Membership: |
|

aub
Genomic Map