Assay Details
| Gene Symbol: aub | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103674.3 | NP_001097144.1 | 3 - 4 | 1000 | 127 | |
| NM_057386.5 | NP_476734.1 | 4 - 5 | 1017 | 127 | ||
| GenBank mRNA | AF145680.1 | - | 4 - 5 | 934 | 127 | |
More Information
Set Membership: |
|

aub
Genomic Map