Assay Details
| Gene Symbol: aub | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103674.3 | NP_001097144.1 | 4 - 5 | 1144 | 68 | |
| NM_057386.5 | NP_476734.1 | 5 - 6 | 1161 | 68 | ||
| GenBank mRNA | AF145680.1 | - | 5 - 6 | 1078 | 68 | |
More Information
Set Membership: |
|

aub
Genomic Map