Assay Details
| Gene Symbol: mts | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298795.1 | NP_001285724.1 | 3 - 4 | 1260 | 65 | |
| NM_001298796.1 | NP_001285725.1 | 3 - 4 | 1117 | 65 | ||
| NM_057457.4 | NP_476805.1 | 3 - 4 | 1117 | 65 | ||
| GenBank mRNA | AY058571.1 | - | 3 - 4 | 1167 | 65 | |
| X55199.1 | - | 1046 | 65 | |||
| X78577.1 | - | 1046 | 65 | |||
More Information
Set Membership: |
|

mts
Genomic Map