Assay Details
| Gene Symbol: Wnt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299322.1 | NP_001286251.1 | 4 - 5 | 1773 | 84 | |
| NM_057462.5 | NP_476810.1 | 4 - 5 | 1778 | 84 | ||
| GenBank mRNA | BT133463.1 | - | 4 - 5 | 1775 | 84 | |
More Information
Set Membership: |
|

Wnt2
Genomic Map