Assay Details
| Gene Symbol: Map60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057604.5 | NP_476952.1 | 1 - 2 | 435 | 94 | |
| GenBank mRNA | AY069313.1 | - | 1 - 2 | 455 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Map60| Gene Symbol: Map60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057604.5 | NP_476952.1 | 1 - 2 | 435 | 94 | |
| GenBank mRNA | AY069313.1 | - | 1 - 2 | 455 | 94 | |
Set Membership: |
|