Assay Details
| Gene Symbol: Taf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057609.6 | NP_476957.1 | 1 | 2052 | 101 | |
| GenBank mRNA | AY051960.1 | - | 1 | 2029 | 101 | |
| U06460.1 | - | 1 | 2033 | 101 | ||
More Information
Set Membership: |
|

Taf5
Genomic Map