Assay Details
| Gene Symbol: dpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299231.1 | NP_001286160.1 | 6 - 7 | 2881 | 99 | |
| NM_057837.5 | NP_477185.1 | 6 - 7 | 2613 | 99 | ||
More Information
Set Membership: |
|

dpa
Genomic Map