Assay Details
| Gene Symbol: ttv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274042.1 | NP_001260971.1 | 9 - 10 | 3096 | 67 | |
| NM_001274043.1 | NP_001260972.1 | 6 - 7 | 1144 | 67 | ||
| NM_001369957.1 | NP_001356963.1 | 9 - 10 | 2787 | 67 | ||
| NM_057883.3 | NP_477231.1 | 9 - 10 | 2787 | 67 | ||
| GenBank mRNA | BT021403.1 | - | 9 - 10 | 2792 | 67 | |
| BT124899.1 | - | 6 - 7 | 1144 | 67 | ||
More Information
Set Membership: |
|

ttv
Genomic Map