Assay Details
| Gene Symbol: eRF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273486.1 | NP_001260415.1 | 1 - 2 | 966 | 57 | |
| NM_001298945.1 | NP_001285874.1 | 1 - 2 | 526 | 57 | ||
| NM_057911.4 | NP_477259.1 | 1 - 2 | 966 | 57 | ||
| GenBank mRNA | AY070979.1 | - | 1 - 2 | 539 | 57 | |
More Information
Set Membership: |
|

eRF3
Genomic Map