Assay Details
| Gene Symbol: eRF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273486.1 | NP_001260415.1 | 4 - 5 | 2017 | 61 | |
| NM_001273487.1 | NP_001260416.1 | 4 - 5 | 1334 | 61 | ||
| NM_001298945.1 | NP_001285874.1 | 4 - 5 | 1577 | 61 | ||
| NM_057911.4 | NP_477259.1 | 4 - 5 | 2017 | 61 | ||
| GenBank mRNA | AY070979.1 | - | 4 - 5 | 1590 | 61 | |
More Information
Set Membership: |
|

eRF3
Genomic Map