Assay Details
| Gene Symbol: Dap160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169557.2 | NP_001163028.1 | 2 - 3 | 495 | 57 | |
| NM_001259189.2 | NP_001246118.1 | 2 - 3 | 495 | 57 | ||
| NM_001273734.1 | NP_001260663.1 | 2 - 3 | 495 | 57 | ||
| NM_001273735.1 | NP_001260664.1 | 2 - 3 | 495 | 57 | ||
| NM_057974.4 | NP_477322.1 | 2 - 3 | 495 | 57 | ||
| NM_057975.4 | NP_477323.1 | 2 - 3 | 495 | 57 | ||
| GenBank mRNA | BT024977.1 | - | 2 - 3 | 57 | ||
More Information
Set Membership: |
|

Dap160
Genomic Map