Assay Details
| Gene Symbol: Eip63F-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080163.4 | NP_524902.2 | 1 - 2 | 394 | 61 | |
| GenBank mRNA | BT088940.1 | - | 1 - 2 | 373 | 61 | |
More Information
Set Membership: |
|

Eip63F-1
Genomic Map