Assay Details
| Gene Symbol: Sam-S | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080184.2 | NP_524923.1 | 4 - 5 | 583 | 66 | |
| NM_164355.3 | NP_722593.1 | 4 - 5 | 512 | 66 | ||
| NM_164361.2 | NP_722599.1 | 4 - 5 | 496 | 66 | ||
| NM_164362.3 | NP_722600.1 | 3 - 4 | 396 | 66 | ||
| GenBank mRNA | BT099838.1 | - | 3 - 4 | 382 | 66 | |
More Information
Set Membership: |
|

Sam-S
Genomic Map