Assay Details
| Gene Symbol: Rbf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080297.3 | NP_525036.2 | 6 - 7 | 2636 | 92 | |
| GenBank mRNA | AY089532.1 | - | 6 - 7 | 2614 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbf| Gene Symbol: Rbf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080297.3 | NP_525036.2 | 6 - 7 | 2636 | 92 | |
| GenBank mRNA | AY089532.1 | - | 6 - 7 | 2614 | 92 | |
Set Membership: |
|