Assay Details
| Gene Symbol: Scgdelta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169168.2 | NP_001162639.2 | 3 - 4 | 1058 | 72 | |
| NM_001169169.2 | NP_001162640.1 | 3 - 4 | 1052 | 72 | ||
| NM_001169170.2 | NP_001162641.1 | 2 - 3 | 1245 | 72 | ||
| NM_080300.3 | NP_525039.1 | 3 - 4 | 1058 | 72 | ||
| GenBank mRNA | AF277393.1 | - | 2 - 3 | 803 | 72 | |
| BT064640.1 | - | 3 - 4 | 1050 | 72 | ||
More Information
Set Membership: |
|

Scgdelta
Genomic Map